Assay Details
Gene Symbol: Nfyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031553.2 | NP_113741.1 | 5 - 6 | 646 | 68 | |
XM_006241196.2 | XP_006241258.1 | 5 - 6 | 664 | 68 | ||
GenBank mRNA | BC089791.1 | - | 5 - 6 | 646 | 68 | |
M55045.1 | - | 4 - 5 | 515 | 68 | ||
M60617.1 | - | 2 - 3 | 319 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
