Assay Details
Gene Symbol: Tmbim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199116.2 | NP_954547.1 | 1 - 2 | 163 | 113 | |
GenBank mRNA | BC060596.1 | - | 1 - 2 | 164 | 113 | |
FQ217672.1 | - | 1 - 2 | 106 | 113 | ||
FQ228009.1 | - | 1 - 2 | 113 | |||
FQ230396.1 | - | 1 - 2 | 128 | 113 | ||
FQ233689.1 | - | 1 - 2 | 157 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
