Assay Details
Gene Symbol: Tmbim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199116.2 | NP_954547.1 | 2 - 3 | 272 | 106 | |
GenBank mRNA | BC060596.1 | - | 2 - 3 | 273 | 106 | |
FQ217672.1 | - | 2 - 3 | 215 | 106 | ||
FQ228009.1 | - | 2 - 3 | 269 | 106 | ||
FQ230396.1 | - | 2 - 3 | 237 | 106 | ||
FQ233689.1 | - | 2 - 3 | 266 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
