Assay Details
Gene Symbol: Ptprq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022925.1 | NP_075214.1 | 2 - 3 | 343 | 70 | |
GenBank mRNA | AF063249.1 | - | 2 - 3 | 343 | 70 |
Gene Symbol: LOC102552564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241319.2 | XP_006241381.1 | 2 - 3 | 347 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
