Assay Details
Gene Symbol: Rtcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207614.1 | NP_997497.1 | 2 - 3 | 172 | 68 | |
GenBank mRNA | AY572415.1 | - | 2 - 3 | 172 | 68 | |
BC078924.1 | - | 2 - 3 | 225 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
