Assay Details
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022539.1 | NP_071984.1 | 7 - 8 | 1049 | 143 | |
GenBank mRNA | BC070892.1 | - | 7 - 8 | 964 | 143 | |
BC091150.1 | - | 7 - 8 | 1063 | 143 | ||
L10652.1 | - | 7 - 8 | 1049 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P065751 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P065753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128417 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P065751 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P065753 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P065751 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P065753 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P065751 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P065753 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15562 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700145442 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97278 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23506 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103909 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24112 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63835 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700145442 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10901926 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901933 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901930 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901925 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901929 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901928 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901934 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901927 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901932 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901935 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901924 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901931 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10901923 | - | - |
Rat Genome 230 2.0 Array | 1398798_at | - | - |
Rat Genome 230 2.0 Array | 1398798_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398798_at | - | - |
Affymetrix HT Rat Focus Array | 1398798_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700019982.rn.1 | TC0700002506.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700019984.rn.1 | TC0700002506.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700019983.rn.1 | TC0700002506.rn.1 | RTA TC |
Rat Expression Set 230 | 1398798_PM_at | - | - |
Rat Expression Set 230 | 1398798_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901923 | - | - |
Rat Genome U34 Set | L10652_g_at | - | - |
Rat Genome U34 Set | L10652_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21315266 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357283 | - | - |
RatRef-12 Expression BeadChip | ILMN_58087 | - | - |
