Assay Details
Gene Symbol: Slc25a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270788.1 | NP_001257717.1 | 5 - 6 | 734 | 87 | |
NM_139100.2 | NP_620800.2 | 5 - 6 | 731 | 87 | ||
GenBank mRNA | BC070918.1 | - | 5 - 6 | 673 | 87 | |
FQ210190.1 | - | 5 - 6 | 685 | 87 | ||
FQ213750.1 | - | 5 - 6 | 690 | 87 | ||
FQ214698.1 | - | 5 - 6 | 688 | 87 | ||
FQ215315.1 | - | 5 - 6 | 688 | 87 | ||
FQ216349.1 | - | 5 - 6 | 687 | 87 | ||
FQ216409.1 | - | 5 - 6 | 695 | 87 | ||
FQ216421.1 | - | 5 - 6 | 688 | 87 | ||
FQ216474.1 | - | 5 - 6 | 696 | 87 | ||
FQ220626.1 | - | 5 - 6 | 672 | 87 | ||
FQ223916.1 | - | 5 - 6 | 688 | 87 | ||
FQ227446.1 | - | 5 - 6 | 685 | 87 | ||
FQ230627.1 | - | 5 - 6 | 687 | 87 | ||
FQ232293.1 | - | 3 - 4 | 490 | 87 | ||
FQ232835.1 | - | 5 - 6 | 681 | 87 | ||
FQ234854.1 | - | 5 - 6 | 685 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
