Assay Details
Gene Symbol: Slc25a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270788.1 | NP_001257717.1 | 3 - 4 | 366 | 126 | |
GenBank mRNA | BC070918.1 | - | 3 - 4 | 305 | 126 | |
FQ214698.1 | - | 3 - 4 | 320 | 126 | ||
FQ215315.1 | - | 3 - 4 | 320 | 126 | ||
FQ215697.1 | - | 3 - 4 | 320 | 126 | ||
FQ215957.1 | - | 3 - 4 | 320 | 126 | ||
FQ216349.1 | - | 3 - 4 | 319 | 126 | ||
FQ216361.1 | - | 3 - 4 | 320 | 126 | ||
FQ216409.1 | - | 3 - 4 | 327 | 126 | ||
FQ216421.1 | - | 3 - 4 | 320 | 126 | ||
FQ216474.1 | - | 3 - 4 | 327 | 126 | ||
FQ217707.1 | - | 3 - 4 | 320 | 126 | ||
FQ223916.1 | - | 3 - 4 | 320 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
