Assay Details
Gene Symbol: Rrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008346.1 | NP_001008347.1 | 2 - 3 | 529 | 144 | |
GenBank mRNA | BC085119.1 | - | 2 - 3 | 529 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
