Assay Details
Gene Symbol: Eif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100662.1 | NP_001094132.1 | 11 - 12 | 1393 | 62 | |
XM_006230226.2 | XP_006230288.1 | 11 - 12 | 1709 | 62 | ||
GenBank mRNA | BC103728.1 | - | 5 - 6 | 583 | 62 | |
BC169011.1 | - | 11 - 12 | 1393 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
