Assay Details
Gene Symbol: Eif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100662.1 | NP_001094132.1 | 18 - 19 | 2616 | 118 | |
XM_006230226.2 | XP_006230288.1 | 18 - 19 | 2932 | 118 | ||
GenBank mRNA | BC103728.1 | - | 12 - 13 | 1806 | 118 | |
BC169011.1 | - | 18 - 19 | 2616 | 118 | ||
FQ230797.1 | - | 5 - 6 | 880 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
