Assay Details
Gene Symbol: Me3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108491.2 | NP_001101961.2 | 5 - 6 | 622 | 85 | |
XM_006229666.1 | XP_006229728.1 | 5 - 6 | 622 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
