Assay Details
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012469.1 | NP_001012487.1 | 5 - 6 | 858 | 77 | |
XM_006230261.2 | XP_006230323.1 | 4 - 5 | 895 | 77 | ||
XM_006230262.2 | XP_006230324.1 | 5 - 6 | 871 | 77 | ||
GenBank mRNA | BC089857.1 | - | 5 - 6 | 858 | 7381492 | 77 |
FQ226709.1 | - | 4 - 5 | 729 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
