Assay Details
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107540.2 | NP_001101010.1 | 3 - 4 | 244 | 104 | |
XM_006229878.2 | XP_006229940.1 | 3 - 4 | 469 | 104 | ||
XM_006229879.2 | XP_006229941.1 | 4 - 5 | 587 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
