Assay Details
Gene Symbol: Pold3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024750.1 | NP_001019921.1 | 3 - 4 | 310 | 79 | |
GenBank mRNA | BC098010.1 | - | 3 - 4 | 310 | 79 | |
FQ211238.1 | - | 312 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
