Assay Details
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133409.2 | NP_596900.1 | 4 - 5 | 517 | 165 | |
GenBank mRNA | BC062406.1 | - | 4 - 5 | 517 | 165 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
