Assay Details
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133409.2 | NP_596900.1 | 8 - 9 | 917 | 171 | |
GenBank mRNA | AF329194.1 | - | 8 - 9 | 854 | 171 | |
BC062406.1 | - | 8 - 9 | 917 | 171 | ||
FQ233927.1 | - | 2 - 3 | 244 | 171 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
