Assay Details
Gene Symbol: Psma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017278.1 | NP_058974.1 | 5 - 6 | 430 | 62 | |
GenBank mRNA | BC062233.1 | - | 5 - 6 | 427 | 62 | |
D90265.1 | - | 5 - 6 | 430 | 62 | ||
FQ213606.1 | - | 5 - 6 | 420 | 62 | ||
FQ215179.1 | - | 5 - 6 | 409 | 62 | ||
FQ215378.1 | - | 5 - 6 | 413 | 62 | ||
FQ216107.1 | - | 5 - 6 | 423 | 62 | ||
FQ218243.1 | - | 462 | 62 | |||
FQ220925.1 | - | 5 - 6 | 395 | 62 | ||
FQ222787.1 | - | 5 - 6 | 423 | 62 | ||
FQ223714.1 | - | 5 - 6 | 399 | 62 | ||
FQ225726.1 | - | 5 - 6 | 381 | 62 | ||
FQ229288.1 | - | 5 - 6 | 457 | 62 | ||
M29859.1 | - | 5 - 6 | 430 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
