Assay Details
Gene Symbol: Relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108495.1 | NP_001101965.1 | 4 - 5 | 398 | 72 | |
XM_006229775.2 | XP_006229837.1 | 5 - 6 | 566 | 72 | ||
XM_006229776.2 | XP_006229838.1 | 5 - 6 | 2830 | 72 | ||
XM_006229777.2 | XP_006229839.1 | 5 - 6 | 517 | 72 | ||
XM_006229778.2 | XP_006229840.1 | 5 - 6 | 4918 | 72 | ||
XM_006229780.2 | XP_006229842.1 | 5 - 6 | 566 | 72 | ||
XM_008759763.1 | XP_008757985.1 | 5 - 6 | 5246 | 72 | ||
XM_008759764.1 | XP_008757986.1 | 5 - 6 | 5171 | 72 | ||
XM_008759765.1 | XP_008757987.1 | 5 - 6 | 5126 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
