Assay Details
Gene Symbol: Prss53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109156.1 | NP_001102626.1 | 7 - 8 | 1194 | 61 | |
XM_006230322.2 | XP_006230384.1 | 7 - 8 | 1835 | 61 | ||
XM_006230324.2 | XP_006230386.1 | 7 - 8 | 1865 | 61 | ||
XM_008759914.1 | XP_008758136.1 | 6 - 7 | 1983 | 61 | ||
GenBank mRNA | BC161977.1 | - | 6 - 7 | 1304 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
