Assay Details
Gene Symbol: Pdilt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013902.1 | NP_001013924.1 | 10 - 11 | 1625 | 84 | |
XM_008759714.1 | XP_008757936.1 | 9 - 10 | 1444 | 84 | ||
GenBank mRNA | BC083897.1 | - | 10 - 11 | 1625 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
