Assay Details
Gene Symbol: Pdilt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013902.1 | NP_001013924.1 | 1 - 2 | 211 | 85 | |
XM_008759714.1 | XP_008757936.1 | 1 - 2 | 228 | 85 | ||
GenBank mRNA | BC083897.1 | - | 1 - 2 | 211 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083897.1, NM_001013902.1, XM_008759714.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
