Assay Details
Gene Symbol: Tgfb1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191840.1 | NP_001178769.1 | 3 - 4 | 206 | 126 | |
XM_006230349.2 | XP_006230411.1 | 3 - 4 | 1056 | 126 | ||
XM_008759921.1 | XP_008758143.1 | 3 - 4 | 267 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
