Assay Details
Gene Symbol: Slc14a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019347.2 | NP_062220.2 | 3 - 4 | 367 | 125 | |
NM_177962.3 | NP_808877.2 | 3 - 4 | 367 | 125 | ||
GenBank mRNA | AF031642.1 | - | 3 - 4 | 367 | 125 | |
AF041788.1 | - | 3 - 4 | 367 | 125 | ||
AF042167.1 | - | 3 - 4 | 367 | 125 | ||
U77971.1 | - | 3 - 4 | 367 | 125 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF031642.1, AF041788.1, AF042167.1, NM_019347.2, NM_177962.3, U77971.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
