Assay Details
Gene Symbol: Slc14a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110270.1 | NP_001103740.1 | 8 - 9 | 1733 | 64 | |
NM_019347.2 | NP_062220.2 | 19 - 20 | 2777 | 64 | ||
GenBank mRNA | AF042167.1 | - | 10 - 11 | 1388 | 64 | |
U09957.1 | - | 8 - 9 | 1732 | 64 | ||
U77971.1 | - | 19 - 20 | 2777 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
