Assay Details
Gene Symbol: Cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053527.1 | NP_445979.1 | 9 - 10 | 1341 | 82 | |
GenBank mRNA | AF000578.2 | - | 9 - 10 | 1341 | 82 | |
BC161839.1 | - | 9 - 10 | 1305 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
