Assay Details
Gene Symbol: Hsp90ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004082.3 | NP_001004082.3 | 1 - 2 | 46 | 68 | |
GenBank mRNA | AY695392.1 | - | 1 - 2 | 46 | 68 | |
AY695393.1 | - | 1 - 2 | 34 | 68 | ||
BC082009.1 | - | 1 - 2 | 72 | 68 | ||
FQ225238.1 | - | 1 - 2 | 98 | 68 | ||
FQ225402.1 | - | 1 - 2 | 98 | 68 | ||
FQ227008.1 | - | 1 - 2 | 98 | 68 | ||
S45392.1 | - | 1 - 2 | 78 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY695392.1, AY695393.1, BC082009.1, NM_001004082.3, S45392.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
