Assay Details
Gene Symbol: Cenpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014215.1 | NP_001014237.1 | 4 - 5 | 463 | 153 | |
GenBank mRNA | BC082113.1 | - | 4 - 5 | 463 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
