Assay Details
Gene Symbol: Mrpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034136.1 | NP_001029308.1 | 5 - 6 | 736 | 90 | |
GenBank mRNA | BC100081.1 | - | 5 - 6 | 736 | 90 | |
FQ214924.1 | - | 5 - 6 | 769 | 90 | ||
FQ229593.1 | - | 5 - 6 | 770 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
