Assay Details
Gene Symbol: Clic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053603.2 | NP_446055.1 | 3 - 4 | 557 | 88 | |
GenBank mRNA | AF323174.1 | - | 3 - 4 | 565 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
