Assay Details
Gene Symbol: Runx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278483.1 | NP_001265412.1 | 2 - 3 | 259 | 116 | |
XM_006244549.2 | XP_006244611.1 | 1 - 2 | 460 | 116 | ||
XM_006244550.2 | XP_006244612.1 | 2 - 3 | 471 | 116 | ||
XM_006244551.2 | XP_006244613.1 | 2 - 3 | 438 | 116 | ||
GenBank mRNA | AB115746.1 | - | 1 - 2 | 59 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
