Assay Details
Gene Symbol: Runx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278483.1 | NP_001265412.1 | 6 - 7 | 1085 | 149 | |
XM_006244549.2 | XP_006244611.1 | 5 - 6 | 1286 | 149 | ||
XM_006244550.2 | XP_006244612.1 | 6 - 7 | 1297 | 149 | ||
XM_006244551.2 | XP_006244613.1 | 6 - 7 | 1264 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
