Assay Details
Gene Symbol: Mut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001067239.5 | XP_001067239.3 | 3 - 4 | 1004 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
