Assay Details
Gene Symbol: Vsig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037784.1 | NP_001032873.1 | 3 - 4 | 562 | 137 | |
XM_006234243.2 | XP_006234305.1 | 4 - 5 | 2885 | 137 | ||
XM_006234244.2 | XP_006234306.1 | 3 - 4 | 2776 | 137 | ||
XM_006234245.2 | XP_006234307.1 | 2 - 3 | 3213 | 137 | ||
XM_006234246.2 | XP_006234308.1 | 2 - 3 | 1464 | 137 | ||
GenBank mRNA | BC098943.1 | - | 3 - 4 | 562 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
