Assay Details
Gene Symbol: Mtch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106488.2 | NP_001099958.2 | 4 - 5 | 377 | 107 | |
GenBank mRNA | BC158687.1 | - | 4 - 5 | 328 | 107 | |
DY311538.1 | - | 328 | 107 | |||
EV779017.1 | - | 398 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
