Assay Details
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234763.1 | XP_006234825.1 | 1 - 2 | 154 | 75 | |
XM_008762100.1 | XP_008760322.1 | 1 - 2 | 154 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006234763.1, XM_008762100.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
