Assay Details
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004265.2 | NP_001004265.2 | 3 - 4 | 947 | 69 | |
XM_006234763.1 | XP_006234825.1 | 4 - 5 | 959 | 69 | ||
XM_006234764.2 | XP_006234826.1 | 4 - 5 | 885 | 69 | ||
XM_008762100.1 | XP_008760322.1 | 4 - 5 | 959 | 69 | ||
GenBank mRNA | AB154834.1 | - | 3 - 4 | 720 | 69 | |
BC097253.1 | - | 3 - 4 | 947 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
