Assay Details
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004265.2 | NP_001004265.2 | 4 - 5 | 1118 | 71 | |
XM_006234763.1 | XP_006234825.1 | 5 - 6 | 1130 | 71 | ||
XM_006234764.2 | XP_006234826.1 | 5 - 6 | 1056 | 71 | ||
XM_008762100.1 | XP_008760322.1 | 5 - 6 | 1130 | 71 | ||
GenBank mRNA | BC097253.1 | - | 4 - 5 | 1118 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
