Assay Details
Gene Symbol: Itpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107774.1 | NP_001101244.1 | 4 - 5 | 323 | 83 | |
GenBank mRNA | FQ230286.1 | - | 4 - 5 | 286 | 83 | |
FQ231907.1 | - | 4 - 5 | 286 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
