Assay Details
Gene Symbol: Itpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107774.1 | NP_001101244.1 | 7 - 8 | 556 | 75 | |
GenBank mRNA | FQ230286.1 | - | 7 - 8 | 519 | 75 | |
FQ231907.1 | - | 7 - 8 | 519 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
