Assay Details
Gene Symbol: Ehf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106493.2 | NP_001099963.1 | 4 - 5 | 539 | 100 | |
XM_006234639.2 | XP_006234701.1 | 4 - 5 | 3038 | 100 | ||
XM_006234641.2 | XP_006234703.1 | 4 - 5 | 613 | 100 | ||
GenBank mRNA | BC166742.1 | - | 4 - 5 | 539 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
