Assay Details
Gene Symbol: Ehf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106493.2 | NP_001099963.1 | 6 - 7 | 676 | 83 | |
XM_006234639.2 | XP_006234701.1 | 6 - 7 | 3175 | 83 | ||
GenBank mRNA | BC166742.1 | - | 6 - 7 | 676 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
