Assay Details
Gene Symbol: Ehf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106493.2 | NP_001099963.1 | 1 - 2 | 131 | 79 | |
XM_006234641.2 | XP_006234703.1 | 1 - 2 | 205 | 79 | ||
GenBank mRNA | BC166742.1 | - | 1 - 2 | 131 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC166742.1, NM_001106493.2, XM_006234641.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
