Assay Details
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012184.1 | NP_001012184.1 | 7 - 8 | 1187 | 77 | |
XM_006234234.2 | XP_006234296.1 | 7 - 8 | 944 | 77 | ||
XM_006234235.2 | XP_006234297.1 | 7 - 8 | 1237 | 77 | ||
XM_006234236.2 | XP_006234298.1 | 7 - 8 | 1036 | 77 | ||
XM_006234237.2 | XP_006234299.1 | 7 - 8 | 1796 | 77 | ||
XM_006234238.2 | XP_006234300.1 | 7 - 8 | 944 | 77 | ||
XM_006234239.2 | XP_006234301.1 | 8 - 9 | 1434 | 77 | ||
XM_006234241.2 | XP_006234303.1 | 7 - 8 | 1034 | 77 | ||
XM_008761860.1 | XP_008760082.1 | 7 - 8 | 1525 | 77 | ||
GenBank mRNA | BC087094.1 | - | 7 - 8 | 1187 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
