Assay Details
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234797.2 | XP_006234859.1 | 10 - 11 | 3818 | 112 | |
XM_006234798.2 | XP_006234860.1 | 10 - 11 | 4353 | 112 | ||
XM_006234799.2 | XP_006234861.1 | 10 - 11 | 3877 | 112 | ||
XM_006234800.2 | XP_006234862.1 | 10 - 11 | 3818 | 112 | ||
XM_008762120.1 | XP_008760342.1 | 10 - 11 | 4119 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
