Assay Details
Gene Symbol: C1qtnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107745.1 | NP_001101215.1 | 1 - 2 | 209 | 128 | |
GenBank mRNA | BC168928.1 | - | 1 - 2 | 222 | 128 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168928.1, NM_001107745.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
