Assay Details
Gene Symbol: Katnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109645.2 | NP_001103115.1 | 3 - 4 | 243 | 87 | |
XM_006234712.2 | XP_006234774.1 | 3 - 4 | 296 | 87 | ||
GenBank mRNA | BC161951.1 | - | 3 - 4 | 243 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
