Assay Details
Gene Symbol: Katnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109645.2 | NP_001103115.1 | 9 - 10 | 973 | 141 | |
XM_006234712.2 | XP_006234774.1 | 9 - 10 | 1026 | 141 | ||
GenBank mRNA | BC161951.1 | - | 9 - 10 | 973 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P319382 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P122726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162245 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P319382 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P122726 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P319382 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P122726 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P319382 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P122726 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P329015 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P319382 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10838504 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838503 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838500 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838505 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838501 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838499 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838502 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838498 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10838497 | - | - |
Rat Genome 230 2.0 Array | 1392641_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394629_at | - | - |
Rat Genome 230 2.0 Array | 1393580_at | - | - |
Rat Genome 230 2.0 Array | 1394629_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390889_at | - | - |
Rat Genome 230 2.0 Array | 1393580_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390889_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392641_at | - | - |
Affymetrix HT Rat Focus Array | 1392641_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394629_at | - | - |
Affymetrix HT Rat Focus Array | 1392641_at | - | - |
Affymetrix HT Rat Focus Array | 1393580_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394629_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393580_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300009462.rn.1 | TC0300001414.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300009457.rn.1 | TC0300001414.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300009458.rn.1 | TC0300001414.rn.1 | RTA TC |
Rat Expression Set 230 | 1394629_PM_at | - | - |
Rat Expression Set 230 | 1390889_PM_at | - | - |
Rat Expression Set 230 | 1394629_at | - | - |
Rat Expression Set 230 | 1392641_PM_at | - | - |
Rat Expression Set 230 | 1393580_at | - | - |
Rat Expression Set 230 | 1390889_at | - | - |
Rat Expression Set 230 | 1393580_PM_at | - | - |
Rat Expression Set 230 | 1392641_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838497 | - | - |
Rat Genome U34 Set | rc_AI169596_at | - | - |
Rat Genome U34 Set | rc_AI230340_at | - | - |
