Assay Details
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013919.1 | NP_001013941.1 | 4 - 5 | 430 | 78 | |
XM_006234904.2 | XP_006234966.1 | 3 - 4 | 885 | 78 | ||
XM_006234906.2 | XP_006234968.1 | 3 - 4 | 390 | 78 | ||
GenBank mRNA | BC083716.1 | - | 4 - 5 | 430 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
