Assay Details
Gene Symbol: Slc27a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031736.1 | NP_113924.1 | 5 - 6 | 1964 | 76 | |
GenBank mRNA | BC081766.1 | - | 5 - 6 | 1301 | 76 | |
D85100.1 | - | 5 - 6 | 1964 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
